CH4102152286 ChemBridge Corporation

1-(4-bromophenyl)-1-butanol

  • MDL number MFCD00021926
  • Cas Number : 22135-53-1
  • Molecular Weight : 229.1136

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5101703

Safety Information

MSDS https://www.hit2lead.com/sds/5101703_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart