CH4102152296 ChemBridge Corporation

6-bromo-2,3-quinoxalinediol

  • MDL number MFCD00457865
  • Cas Number : 1910-90-3
  • Molecular Weight : 241.0415

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5131629

Safety Information

MSDS https://www.hit2lead.com/building-blocks/5131629
Availability details for
Quantity
  • Availability
  • Add to Cart