CH4102152300 ChemBridge Corporation

2-amino-5-phenyl-4,6-pyrimidinediol

  • MDL number MFCD00748802
  • Cas Number : 154146-07-3
  • Molecular Weight : 203.1974

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5157250

Safety Information

MSDS https://www.hit2lead.com/sds/5157250_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart