CH4102152305 ChemBridge Corporation

2-methyl-8-quinolinol 1-oxide

  • MDL number MFCD00168978
  • Cas Number : 63543-87-3
  • Molecular Weight : 175.184

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5175127

Safety Information

MSDS https://www.hit2lead.com/sds/5175127_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart