CH4102152319 ChemBridge Corporation

3-allyl-7-hydroxy-4-methyl-2H-chromen-2-one

  • MDL number MFCD00488351
  • Cas Number : 26481-13-0
  • Molecular Weight : 216.2326

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5223960

Safety Information

MSDS https://www.hit2lead.com/sds/5223960_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart