CH4102152325 ChemBridge Corporation

3-chloro-N-(4-methoxybenzyl)propanamide

  • MDL number MFCD00455271
  • Cas Number : 2364-76-3
  • Molecular Weight : 227.6874

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5238058

Safety Information

MSDS https://www.hit2lead.com/sds/5238058_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart