CH4102152331 ChemBridge Corporation

1-(4-bromobenzoyl)piperidine

  • MDL number MFCD00447936
  • Cas Number : 98612-93-2
  • Molecular Weight : 268.1497

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5264381

Safety Information

MSDS https://www.hit2lead.com/sds/5264381_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart