CH4102152334 ChemBridge Corporation

4-(1-azepanyl)-4-oxobutanoic acid

  • MDL number MFCD00594274
  • Cas Number : 154740-93-9
  • Molecular Weight : 199.2469

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5282144

Safety Information

MSDS https://www.hit2lead.com/building-blocks/5282144
Availability details for
Quantity
  • Availability
  • Add to Cart