CH4102152337 ChemBridge Corporation

3-cyclohexyl-1H-1,2,4-triazol-5-amine

  • MDL number MFCD00755379
  • Cas Number : 90111-72-1
  • Molecular Weight : 166.2236

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5302581

Safety Information

MSDS https://www.hit2lead.com/building-blocks/5302581
Availability details for
Quantity
  • Availability
  • Add to Cart