CH4102152342 ChemBridge Corporation

2,6-dimethyl-4-(1-piperidinylmethyl)phenol

  • MDL number MFCD02224567
  • Cas Number : 42900-97-0
  • Molecular Weight : 219.3226

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5337566

Safety Information

MSDS https://www.hit2lead.com/sds/5337566_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart