CH4102152347 ChemBridge Corporation

3-chloro-N-(2-furylmethyl)propanamide

  • MDL number MFCD02973608
  • Cas Number : 64017-87-4
  • Molecular Weight : 187.6235

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5344117

Safety Information

MSDS https://www.hit2lead.com/sds/5344117_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart