CH4102152353 ChemBridge Corporation

2-bromo-N,N-dimethylbenzamide

  • MDL number MFCD00463635
  • Cas Number : 54616-47-6
  • Molecular Weight : 228.0858

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5346231

Safety Information

MSDS https://www.hit2lead.com/building-blocks/5346231
Availability details for
Quantity
  • Availability
  • Add to Cart