CH4102152356 ChemBridge Corporation

3-chloro-N-(2,4-dimethoxyphenyl)propanamide

  • MDL number MFCD02973794
  • Cas Number : 349097-71-8
  • Molecular Weight : 243.6868

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5346762

Safety Information

MSDS https://www.hit2lead.com/sds/5346762_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart