CH4102152365 ChemBridge Corporation

3-chloro-N-(2-methylphenyl)propanamide

  • MDL number MFCD02286135
  • Cas Number : 19422-76-5
  • Molecular Weight : 197.6614

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5349373

Safety Information

MSDS https://www.hit2lead.com/sds/5349373_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart