CH4102152369 ChemBridge Corporation

3-chloro-N-(4-ethylphenyl)propanamide

  • MDL number MFCD01353850
  • Cas Number : 20330-92-1
  • Molecular Weight : 211.688

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5350963

Safety Information

MSDS https://www.hit2lead.com/building-blocks/5350963
Availability details for
Quantity
  • Availability
  • Add to Cart