CH4102152400 ChemBridge Corporation

2-(5-phenyl-2H-tetrazol-2-yl)ethanol

  • MDL number MFCD00501885
  • Cas Number : 93742-43-9
  • Molecular Weight : 190.2019

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5380951

Safety Information

MSDS https://www.hit2lead.com/sds/5380951_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart