CH4102152404 ChemBridge Corporation

methyl 4-acetyl-5-methyl-2-furoate

  • MDL number MFCD00463178
  • Cas Number : 29172-10-9
  • Molecular Weight : 182.1733

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5404454

Safety Information

MSDS https://www.hit2lead.com/sds/5404454_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart