CH4102152427 ChemBridge Corporation

4-[(3-hydroxyphenyl)amino]-4-oxobutanoic acid

  • MDL number MFCD00761518
  • Cas Number : 16141-43-8
  • Molecular Weight : 209.1986

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5539774

Safety Information

MSDS https://www.hit2lead.com/sds/5539774_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart