CH4102152439 ChemBridge Corporation

3-(4-fluorophenyl)-3-phenylpropanoic acid

  • MDL number MFCD01897573
  • Cas Number : 362-86-7
  • Molecular Weight : 244.2609

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5553673

Safety Information

MSDS https://www.hit2lead.com/sds/5553673_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart