CH4102152441 ChemBridge Corporation

ethyl 3-[(chloroacetyl)amino]benzoate

  • MDL number MFCD00711338
  • Cas Number : 58915-19-8
  • Molecular Weight : 241.6709

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5554900

Safety Information

MSDS https://www.hit2lead.com/sds/5554900_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart