CH4102152450 ChemBridge Corporation

3-chloro-N-(3,4-difluorophenyl)propanamide

  • MDL number MFCD01213654
  • Cas Number : 132669-28-4
  • Molecular Weight : 219.6157

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5565565

Safety Information

MSDS https://www.hit2lead.com/sds/5565565_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart