CH4102152455 ChemBridge Corporation

N-1,3-benzothiazol-2-yl-3-chloropropanamide

  • MDL number MFCD01213665
  • Cas Number : 2877-36-3
  • Molecular Weight : 240.7093

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5568709

Safety Information

MSDS https://www.hit2lead.com/sds/5568709_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart