CH4102152466 ChemBridge Corporation

4-(4-morpholinylcarbonyl)phenol

  • MDL number MFCD00185859
  • Cas Number : 18137-25-2
  • Molecular Weight : 207.2258

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5581642

Safety Information

MSDS https://www.hit2lead.com/sds/5581642_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart