CH4102152474 ChemBridge Corporation

ethyl 2-[(chloroacetyl)amino]benzoate

  • MDL number MFCD00018892
  • Cas Number : 6307-66-0
  • Molecular Weight : 241.6709

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5618917

Safety Information

MSDS https://www.hit2lead.com/sds/5618917_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart