CH4102152477 ChemBridge Corporation

2-chloro-N-[4-(diethylamino)phenyl]acetamide

  • MDL number MFCD00791331
  • Cas Number : 125983-31-5
  • Molecular Weight : 240.7292

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5622066

Safety Information

MSDS https://www.hit2lead.com/sds/5622066_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart