CH4102152478 ChemBridge Corporation

N-(3-acetylphenyl)propanamide

  • MDL number MFCD00791263
  • Cas Number : 39569-28-3
  • Molecular Weight : 191.2264

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5623290

Safety Information

MSDS https://www.hit2lead.com/sds/5623290_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart