CH4102152481 ChemBridge Corporation

N~1~-(diphenylmethyl)glycinamide

  • MDL number MFCD01475934
  • Cas Number : 63628-88-6
  • Molecular Weight : 240.3003

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5633667

Safety Information

MSDS https://www.hit2lead.com/sds/5633667_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart