CH4102152494 ChemBridge Corporation

methyl 3-[(3-chloropropanoyl)amino]-2-thiophenecarboxylate

  • MDL number MFCD01340734
  • Cas Number : 549478-39-9
  • Molecular Weight : 247.6986

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5695179

Safety Information

MSDS https://www.hit2lead.com/sds/5695179_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart