CH4102152503 ChemBridge Corporation

4-amino-N-isobutyl-1,2,5-oxadiazole-3-carboxamide

  • MDL number MFCD02932869
  • Cas Number : 405278-58-2
  • Molecular Weight : 184.1958

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5730567

Safety Information

MSDS https://www.hit2lead.com/sds/5730567_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart