CH4102152519 ChemBridge Corporation

3-(1-piperidinylcarbonyl)phenol

  • MDL number MFCD01017227
  • Cas Number : 15504-60-6
  • Molecular Weight : 205.253

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5762406

Safety Information

MSDS https://www.hit2lead.com/sds/5762406_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart