CH4102152522 ChemBridge Corporation

2-(allylamino)benzoic acid

  • MDL number MFCD02131352
  • Cas Number : 57397-97-4
  • Molecular Weight : 177.1998

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5772307

Safety Information

MSDS https://www.hit2lead.com/sds/5772307_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart