CH4102152542 ChemBridge Corporation

7-isopropoxy-2-oxo-2H-chromene-3-carboxylic acid

  • MDL number MFCD08691747
  • Cas Number : 914203-21-7
  • Molecular Weight : 248.2314

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5883364

Safety Information

MSDS https://www.hit2lead.com/sds/5883364_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart