CH4102152543 ChemBridge Corporation

N-(4-aminophenyl)-5-methyl-3-furamide

  • MDL number MFCD00448335
  • Cas Number : 887358-45-4
  • Molecular Weight : 216.2359

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5924772

Safety Information

MSDS https://www.hit2lead.com/sds/5924772_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart