CH4102152544 ChemBridge Corporation

N-(4-aminophenyl)-5-methyl-2-furamide

  • MDL number MFCD00448324
  • Cas Number : 727387-19-1
  • Molecular Weight : 216.2359

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5925667

Safety Information

MSDS https://www.hit2lead.com/sds/5925667_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart