CH4102152545 ChemBridge Corporation

4-[(butylthio)methyl]-5-methyl-2-furoic acid

  • MDL number MFCD00475551
  • Cas Number : 462075-93-0
  • Molecular Weight : 228.3079

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5925680

Safety Information

MSDS https://www.hit2lead.com/sds/5925680_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart