CH4102152559 ChemBridge Corporation

3-(butyrylamino)benzoic acid

  • MDL number MFCD00448227
  • Cas Number : 76209-00-2
  • Molecular Weight : 207.2258

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5941601

Safety Information

MSDS https://www.hit2lead.com/building-blocks/5941601
Availability details for
Quantity
  • Availability
  • Add to Cart