CH4102152574 ChemBridge Corporation

3-chloro-N-(2-methoxy-5-methylphenyl)propanamide

  • MDL number MFCD02860103
  • Cas Number : 349122-20-9
  • Molecular Weight : 227.6874

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6028814

Safety Information

MSDS https://www.hit2lead.com/sds/6028814_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart