CH4102152578 ChemBridge Corporation

5-(2-methylphenyl)-4H-1,2,4-triazol-3-amine

  • MDL number MFCD00475870
  • Cas Number : 59301-23-4
  • Molecular Weight : 174.2025

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6044848

Safety Information

MSDS https://www.hit2lead.com/sds/6044848_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart