CH4102152589 ChemBridge Corporation

N-(2-amino-4-methylphenyl)-2-methylbenzamide

  • MDL number MFCD02031950
  • Cas Number : 402577-84-8
  • Molecular Weight : 240.3003

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6085589

Safety Information

MSDS https://www.hit2lead.com/sds/6085589_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart