CH4102152590 ChemBridge Corporation

N-(2-furylmethyl)-3-oxobutanamide

  • MDL number MFCD00457343
  • Cas Number : 392721-34-5
  • Molecular Weight : 181.1885

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6086007

Safety Information

MSDS https://www.hit2lead.com/sds/6086007_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart