CH4102152594 ChemBridge Corporation

2-(butyrylamino)-4-chlorobenzoic acid

  • MDL number MFCD02032437
  • Cas Number : 777877-45-9
  • Molecular Weight : 241.6709

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6088842

Safety Information

MSDS https://www.hit2lead.com/sds/6088842_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart