CH4102152595 ChemBridge Corporation

N-(2-amino-4-methylphenyl)pentanamide

  • MDL number MFCD02031952
  • Cas Number : 133085-62-8
  • Molecular Weight : 206.2841

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6089887

Safety Information

MSDS https://www.hit2lead.com/sds/6089887_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart