CH4102152598 ChemBridge Corporation

N-[4-(dimethylamino)phenyl]-3-oxobutanamide

  • MDL number MFCD01001277
  • Cas Number : 38659-86-8
  • Molecular Weight : 220.2676

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6094311

Safety Information

MSDS https://www.hit2lead.com/sds/6094311_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart