CH4102152601 ChemBridge Corporation

2-[(butylamino)carbonyl]cyclohexanecarboxylic acid

  • MDL number MFCD01355608
  • Cas Number : 438613-40-2
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://www.hit2lead.com/building-blocks/6107143

Safety Information

msds https://www.hit2lead.com/building-blocks/6107143
Availability details for
Quantity
  • Availability
  • Add to Cart