CH4102152622 ChemBridge Corporation

N-(5-amino-2-chlorophenyl)-2-furamide

  • MDL number MFCD03724054
  • Cas Number : 505066-17-1
  • Molecular Weight : 236.6544

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6365088

Safety Information

MSDS https://www.hit2lead.com/sds/6365088_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart