CH4102152805 ChemBridge Corporation

3-[4-(aminocarbonyl)-1-piperidinyl]propanoic acid

  • MDL number MFCD03161182
  • Cas Number : 915922-36-0
  • Molecular Weight : 200.2349

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6716342

Safety Information

MSDS https://www.hit2lead.com/sds/6716342_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart