CH4102152827 ChemBridge Corporation

3-amino-N-ethyl-N-phenylbenzamide

  • MDL number MFCD08691808
  • Cas Number : 875837-45-9
  • Molecular Weight : 240.3003

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7991742

Safety Information

MSDS https://www.hit2lead.com/building-blocks/7991742
Availability details for
Quantity
  • Availability
  • Add to Cart