CH4102152840 ChemBridge Corporation

2-[3-(4-methylphenyl)-1,2,4-oxadiazol-5-yl]aniline

  • MDL number MFCD08691851
  • Cas Number : 58589-02-9
  • Molecular Weight : 251.2832

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9028710

Safety Information

MSDS https://www.hit2lead.com/sds/9028710_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart