CH4102152852 ChemBridge Corporation

2-(2-chloroethoxy)-1,3-dimethoxybenzene

  • MDL number MFCD08691875
  • Cas Number : 24251-50-1
  • Molecular Weight : 216.6614

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070502

Safety Information

MSDS https://www.hit2lead.com/sds/9070502_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart