CH4102152860 ChemBridge Corporation

N-ethyl-2-(2-methoxyphenoxy)ethanamine

  • MDL number MFCD08691888
  • Cas Number : 4766-09-0
  • Molecular Weight : 195.2582

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9070636

Safety Information

MSDS https://www.hit2lead.com/sds/9070636_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart